Bromocycloheptane, 97%
Bromocycloheptane, 97%
Bromocycloheptane, 97%
Thermo Scientific Chemicals

Bromocycloheptane, 97%

CAS: 2404-35-5 | C7H13Br | 177.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number B23110.06
also known as B23110-06
Price (USD)
29.90
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
29.90
Each
Chemical Identifiers
CAS2404-35-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5020-1.5070 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only