Methyl p-toluate, 99%
Methyl p-toluate, 99%
Methyl p-toluate, 99%
Thermo Scientific Chemicals

Methyl p-toluate, 99%

CAS: 99-75-2 | C9H10O2 | 150.177 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B23111.22
also known as B23111-22
Price (USD)
32.65
Special offer
Online exclusive
Ends: 30-Jun-2026
38.60
Save 5.95 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
32.65
Special offer
Online exclusive
Ends: 30-Jun-2026
38.60
Save 5.95 (15%)
Each
Chemical Identifiers
CAS99-75-2
IUPAC Namemethyl 4-methylbenzoate
Molecular FormulaC9H10O2
InChI KeyQSSJZLPUHJDYKF-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)30.0-37.0°C
Appearance (Color)Clear colorless to pale yellow as liquid or white to pale yellow as solid
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only