Thermo Scientific Chemicals

Di-n-propyl sulfide, 98+%, Thermo Scientific Chemicals

Catalog number: B23155.36
500 g, Each
Thermo Scientific Chemicals

Di-n-propyl sulfide, 98+%, Thermo Scientific Chemicals

Catalog number: B23155.36
500 g, Each
Quantity
Catalog number: B23155.36
also known as B23155-36
Price (USD)
Price: 860.00
Online price: 731.65
Your price:
Quantity
-

Chemical Identifiers

CAS
111-47-7
IUPAC Name
1-(propylsulfanyl)propane
Molecular Formula
C6H14S
InChI Key
ZERULLAPCVRMCO-UHFFFAOYSA-N
SMILES
CCCSCCC
Appearance (Color)
Clear colorless
Assay (GC)
≥98.0%
Refractive Index
1.4470-1.4510 @ 20?C
Form
Liquid

Description

Di-n-propyl sulfide is used in organic synthesis and used as a daily fragrance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-propyl sulfide is used in organic synthesis and used as a daily fragrance.

Solubility
Soluble in water 351mg/L @ 25°C.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text