2,4-Dimethylthiophenol, 95%
2,4-Dimethylthiophenol, 95%
2,4-Dimethylthiophenol, 95%
2,4-Dimethylthiophenol, 95%
Thermo Scientific Chemicals

2,4-Dimethylthiophenol, 95%

CAS: 13616-82-5 | C8H10S | 138.228 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B23161.14
also known as B23161-14
Price (USD)
252.00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
252.00
Each
Add to cart
Chemical Identifiers
CAS13616-82-5
IUPAC Name2,4-dimethylbenzene-1-thiol
Molecular FormulaC8H10S
InChI KeyAMNLXDDJGGTIPL-UHFFFAOYSA-N
SMILESCC1=CC=C(S)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5670-1.5730 @ 20°C
Assay (GC)≥94.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only