3,4-Dimethylthiophenol, 98%
3,4-Dimethylthiophenol, 98%
3,4-Dimethylthiophenol, 98%
3,4-Dimethylthiophenol, 98%
Thermo Scientific Chemicals

3,4-Dimethylthiophenol, 98%

CAS: 18800-53-8 | C8H10S | 138.228 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B23168.14
also known as B23168-14
Price (USD)
542.65
Online Exclusive
603.00
Save 60.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
542.65
Online Exclusive
603.00
Save 60.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS18800-53-8
IUPAC Name(3,4-dimethylphenyl)sulfanide
Molecular FormulaC8H9S
InChI KeyIDKCKPBAFOIONK-UHFFFAOYSA-M
SMILESCC1=CC=C([S-])C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5725-1.5765 @ 20?C
Appearance (Color)Clear colorless to pale green
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only