3-(Methylthio)aniline, 97%
3-(Methylthio)aniline, 97%
3-(Methylthio)aniline, 97%
Thermo Scientific Chemicals

3-(Methylthio)aniline, 97%

CAS: 1783-81-9 | C7H9NS | 139.216 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B23184.06
also known as B23184-06
Price (USD)
106.65
Special offer
Online exclusive
Ends: 30-Jun-2026
125.00
Save 18.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
106.65
Special offer
Online exclusive
Ends: 30-Jun-2026
125.00
Save 18.35 (15%)
Each
Chemical Identifiers
CAS1783-81-9
IUPAC Name3-(methylsulfanyl)aniline
Molecular FormulaC7H9NS
InChI KeyKCHLDNLIJVSRPK-UHFFFAOYSA-N
SMILESCSC1=CC=CC(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.6405-1.6455 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only