Triacetoxy(ethyl)silane, 96%
Triacetoxy(ethyl)silane, 96%
Triacetoxy(ethyl)silane, 96%
Thermo Scientific Chemicals

Triacetoxy(ethyl)silane, 96%

CAS: 17689-77-9 | C8H14O6Si | 234.279 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B23187.22
also known as B23187-22
Price (USD)
44.65
Online Exclusive
49.80
Save 5.15 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
44.65
Online Exclusive
49.80
Save 5.15 (10%)
Each
Chemical Identifiers
CAS17689-77-9
IUPAC Namebis(acetyloxy)(ethyl)silyl acetate
Molecular FormulaC8H14O6Si
InChI KeyKXJLGCBCRCSXQF-UHFFFAOYSA-N
SMILESCC[Si](OC(C)=O)(OC(C)=O)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
Identification (FTIR)Conforms
Refractive Index1.4100-1.4160 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
Triacetoxy(ethyl)silane is used in sealants, the substance is used as a cross-linking agent and adhesion promoter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triacetoxy(ethyl)silane is used in sealants, the substance is used as a cross-linking agent and adhesion promoter.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture.
RUO – Research Use Only

General References:

  1. Simon Mallam; Anne-Marie Hecht; Erik Geissler and Pierre Pruvost. Structure of swollen polydimethyl siloxane gels. J. Chem. Phys. 1989, 91 (10), 6447.
  2. H. Leidheiser Jr.; M. De Costa; R. D. Granata. Corrosion Behavior of Steel Pre-Treated with Silanes. Corrosion Behavior of Steel Pre-Treated with Silanes. 1987, 43 (6), 382-387.