Triacetoxy(ethyl)silane is used in sealants, the substance is used as a cross-linking agent and adhesion promoter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Triacetoxy(ethyl)silane is used in sealants, the substance is used as a cross-linking agent and adhesion promoter.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture.
RUO – Research Use Only
General References:
- Simon Mallam; Anne-Marie Hecht; Erik Geissler and Pierre Pruvost. Structure of swollen polydimethyl siloxane gels. J. Chem. Phys. 1989, 91 (10), 6447.
- H. Leidheiser Jr.; M. De Costa; R. D. Granata. Corrosion Behavior of Steel Pre-Treated with Silanes. Corrosion Behavior of Steel Pre-Treated with Silanes. 1987, 43 (6), 382-387.