2-Amino-4-(2-naphthyl)thiazole, 98%
2-Amino-4-(2-naphthyl)thiazole, 98%
2-Amino-4-(2-naphthyl)thiazole, 98%
Thermo Scientific Chemicals

2-Amino-4-(2-naphthyl)thiazole, 98%

CAS: 21331-43-1 | C13H10N2S | 226.297 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B23223.06
also known as B23223-06
Price (USD)
84.65
Online Exclusive
94.10
Save 9.45 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
84.65
Online Exclusive
94.10
Save 9.45 (10%)
Each
Chemical Identifiers
CAS21331-43-1
IUPAC Name4-(naphthalen-2-yl)-1,3-thiazol-2-amine
Molecular FormulaC13H10N2S
InChI KeyGWDNDNTTXIIXRS-UHFFFAOYSA-N
SMILESNC1=NC(=CS1)C1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream
Melting Point (clear melt)150.0-157.0?C
FormPowder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only