Fluorene-9-carboxylic acid, 97%
Fluorene-9-carboxylic acid, 97%
Fluorene-9-carboxylic acid, 97%
Fluorene-9-carboxylic acid, 97%
Thermo Scientific Chemicals

Fluorene-9-carboxylic acid, 97%

CAS: 1989-33-9 | C14H10O2 | 210.232 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B23224.14
also known as B23224-14
Price (USD)
112.65
Special offer
Online exclusive
Ends: 30-Jun-2026
132.00
Save 19.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
112.65
Special offer
Online exclusive
Ends: 30-Jun-2026
132.00
Save 19.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS1989-33-9
IUPAC Name9H-fluorene-9-carboxylic acid
Molecular FormulaC14H10O2
InChI KeyDNVJGJUGFFYUPT-UHFFFAOYSA-N
SMILESOC(=O)C1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (HPLC)≥96.0%
Melting Point (clear melt)224.0-234.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only