Methyl Isonicotinate induces increased walking and take-off behavior in western flower thrips, Frankliniella occidentalis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl Isonicotinate induces increased walking and take-off behavior in western flower thrips, Frankliniella occidentalis.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
General References:
- Ronald Breslow; Michael Brandl; Jurgen Hunger; Nicholas Turro; Karen Cassidy; Karsten Krogh-Jespersen; John D. Westbrook. Pyridine complexes of chlorine atoms. J. Am. Chem. Soc. 1987 109 (23), 7204-7206.