2-Ethyl-2-oxazoline, 99%
2-Ethyl-2-oxazoline, 99%
Thermo Scientific™

2-Ethyl-2-oxazoline, 99%

CAS: 10431-98-8 | C5H9NO | 99.13 g/mol
Have Questions?
Catalog number B23287.36
also known as B23287-36
Price (USD)
266.65
Online Exclusive
296.00
Save 29.35 (10%)
Each
Add to cart
Request bulk or custom format
Price (USD)
266.65
Online Exclusive
296.00
Save 29.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS10431-98-8
IUPAC Name2-ethyl-4,5-dihydro-1,3-oxazole
Molecular FormulaC5H9NO
InChI KeyNYEZZYQZRQDLEH-UHFFFAOYSA-N
SMILESCCC1=NCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.4365-1.4395 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
2-ethyl-2-oxazoline is an organic intermediate and monomer that is used in water-soluble polymers, pharmaceuticals, cosmetics, specialty coatings and adhesives.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.