4-Allylanisole, 98%
4-Allylanisole, 98%
4-Allylanisole, 98%
Thermo Scientific Chemicals

4-Allylanisole, 98%

CAS: 140-67-0 | C10H12O | 148.205 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B23311.14
also known as B23311-14
Price (USD)
32.65
Special offer
Online exclusive
Ends: 28-Dec-2026
38.00
Save 5.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
32.65
Special offer
Online exclusive
Ends: 28-Dec-2026
38.00
Save 5.35
Each
Chemical Identifiers
CAS140-67-0
IUPAC Name1-methoxy-4-(prop-2-en-1-yl)benzene
Molecular FormulaC10H12O
InChI KeyZFMSMUAANRJZFM-UHFFFAOYSA-N
SMILESCOC1=CC=C(CC=C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5195-1.5235 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only