2,3-Dimethylquinoxaline, 97%
2,3-Dimethylquinoxaline, 97%
2,3-Dimethylquinoxaline, 97%
Thermo Scientific Chemicals

2,3-Dimethylquinoxaline, 97%

CAS: 2379-55-7 | C10H10N2 | 158.204 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B23313.06
also known as B23313-06
Price (USD)
33.30
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
33.30
Each
Chemical Identifiers
CAS2379-55-7
IUPAC Name2,3-dimethylquinoxaline
Molecular FormulaC10H10N2
InChI KeyFKHNZQFCDGOQGV-UHFFFAOYSA-N
SMILESCC1=C(C)N=C2C=CC=CC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
FormPowder or crystalline powder and/or lumps
Assay (GC)≥96.0%
Melting Point (clear melt)101-107°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only