2-Chlorobenzeneboronic acid, 97%
2-Chlorobenzeneboronic acid, 97%
2-Chlorobenzeneboronic acid, 97%
Thermo Scientific Chemicals

2-Chlorobenzeneboronic acid, 97%

CAS: 3900-89-8 | C6H6BClO2 | 156.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B23324.06
also known as B23324-06
Price (USD)
112.65
Special offer
Online exclusive
Ends: 30-Jun-2026
132.00
Save 19.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
112.65
Special offer
Online exclusive
Ends: 30-Jun-2026
132.00
Save 19.35 (15%)
Each
Chemical Identifiers
CAS3900-89-8
IUPAC Name(2-chlorophenyl)boronic acid
Molecular FormulaC6H6BClO2
InChI KeyRRCMGJCFMJBHQC-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
2-Chlorobenzeneboronic acid is used in the preparation of imidazo[1,2-a]pyridine amides which has tuberculostatic activity. It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzeneboronic acid is used in the preparation of imidazo[1,2-a]pyridine amides which has tuberculostatic activity. It is used as pharmaceutical intermediates.

Solubility
Soluble in DMSO, Methanol.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
Richardson; Marianne L.; Stevens; Malcolm F. G. Structural studies on bioactive compounds. Part 37. Suzuki coupling of diaminopyrimidines: a new synthesis of the antimalarial drug pyrimethamine. Journal of Chemical Research.2002, 2002, (10), 482-484.