2-Ethoxynaphthalene, 99%
2-Ethoxynaphthalene, 99%
2-Ethoxynaphthalene, 99%
Thermo Scientific Chemicals

2-Ethoxynaphthalene, 99%

CAS: 93-18-5 | C12H12O | 172.227 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B23327.14
also known as B23327-14
Price (USD)
20.60
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
20.60
Each
Chemical Identifiers
CAS93-18-5
IUPAC Name2-ethoxynaphthalene
Molecular FormulaC12H12O
InChI KeyGUMOJENFFHZAFP-UHFFFAOYSA-N
SMILESCCOC1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only