3-Nitro-alpha-toluenesulfonyl chloride, 95%
3-Nitro-alpha-toluenesulfonyl chloride, 95%
3-Nitro-alpha-toluenesulfonyl chloride, 95%
Thermo Scientific Chemicals

3-Nitro-alpha-toluenesulfonyl chloride, 95%

CAS: 58032-84-1 | C7H6ClNO4S | 235.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B23333.14
also known as B23333-14
Price (USD)
742.65
Online Exclusive
825.00
Save 82.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
742.65
Online Exclusive
825.00
Save 82.35 (10%)
Each
Chemical Identifiers
CAS58032-84-1
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystalline powder
Melting Point (clear melt)95-103?C
Appearance (Color)White to pale cream or pale yellow
Assay (Titration ex Chloride)≥94.0 to ≤106.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Reacts with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, bases, water/ moisture.
RUO – Research Use Only