3-Hydroxy-2,4,6-triiodobenzoic acid, 97%
3-Hydroxy-2,4,6-triiodobenzoic acid, 97%
3-Hydroxy-2,4,6-triiodobenzoic acid, 97%
Thermo Scientific Chemicals

3-Hydroxy-2,4,6-triiodobenzoic acid, 97%

CAS: 53279-72-4 | C7H3I3O3 | 515.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B23355.14
also known as B23355-14
Price (USD)
294.65
Online Exclusive
327.00
Save 32.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
294.65
Online Exclusive
327.00
Save 32.35 (10%)
Each
Chemical Identifiers
CAS53279-72-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow or cream to pale brown
FormPowder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0% (Dry wt. basis)
Assay (Silylated GC)≥96.0%
Assay (HPLC)≥96.0%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only