2-Methyl-1,3-dithiolane, 99%
2-Methyl-1,3-dithiolane, 99%
2-Methyl-1,3-dithiolane, 99%
Thermo Scientific Chemicals

2-Methyl-1,3-dithiolane, 99%

CAS: 5616-51-3 | C4H8S2 | 120.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B23372.14
also known as B23372-14
Price (USD)
194.65
Special offer
Online exclusive
Ends: 30-Jun-2026
229.00
Save 34.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
194.65
Special offer
Online exclusive
Ends: 30-Jun-2026
229.00
Save 34.35 (15%)
Each
Chemical Identifiers
CAS5616-51-3
IUPAC Name2-methyl-1,3-dithiolane
Molecular FormulaC4H8S2
InChI KeyCARJCVDELAMAEJ-UHFFFAOYSA-N
SMILESCC1SCCS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
FormLiquid
Refractive Index1.5625-1.5655 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only