2,4-Dibromophenol, 98%
2,4-Dibromophenol, 98%
2,4-Dibromophenol, 98%
Thermo Scientific Chemicals

2,4-Dibromophenol, 98%

CAS: 615-58-7 | C6H4Br2O | 251.905 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B23394.14
also known as B23394-14
Price (USD)
47.65
Online Exclusive
52.30
Save 4.65 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
47.65
Online Exclusive
52.30
Save 4.65 (9%)
Each
Chemical Identifiers
CAS615-58-7
IUPAC Name2,4-dibromophenol
Molecular FormulaC6H4Br2O
InChI KeyFAXWFCTVSHEODL-UHFFFAOYSA-N
SMILESOC1=CC=C(Br)C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps or fused solid
Appearance (Color)White to pale cream
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only