3-Methyl-3-buten-1-ol, 97%
3-Methyl-3-buten-1-ol, 97%
3-Methyl-3-buten-1-ol, 97%
3-Methyl-3-buten-1-ol, 97%
Thermo Scientific Chemicals

3-Methyl-3-buten-1-ol, 97%

CAS: 763-32-6 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number B23398.AC
also known as B23398-AC
Price (USD)
26.40
Each
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (USD)
26.40
Each
Add to cart
Chemical Identifiers
CAS763-32-6
IUPAC Name3-methylbut-3-en-1-ol
Molecular FormulaC5H10O
InChI KeyCPJRRXSHAYUTGL-UHFFFAOYSA-N
SMILESCC(=C)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4315-1.4365 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only