1,2-Benzisoxazole, 95%
1,2-Benzisoxazole, 95%
1,2-Benzisoxazole, 95%
Thermo Scientific Chemicals

1,2-Benzisoxazole, 95%

CAS: 271-95-4 | C7H5NO | 119.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B23400.03
also known as B23400-03
Price (USD)
40.50
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
40.50
Each
Chemical Identifiers
CAS271-95-4
IUPAC Name1,2-benzoxazole
Molecular FormulaC7H5NO
InChI KeyKTZQTRPPVKQPFO-UHFFFAOYSA-N
SMILESO1N=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5590-1.5650 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥94.0%
1,2-Benzisoxazole derivative zonisamide is a novel antiepileptic drug and is effective for the treatment of partial seizures. 1,2-Benzisoxazole is a potential substrates of rabbit liver aldehyde oxidase. It is used to produce 5-nitro-benzo[d]isoxazole. It is involved in manufacturing medicines ( antiepileptics and antipsychotics).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water. 4.1 mg/mL in water at 25°C

Notes
Light Sensitive. Store at 4°C. Store in the dark. Incompatible with light and oxidizing agents.
RUO – Research Use Only

General References:

  1. D H Peters; E M Sorkin. Zonisamide. A review of its pharmacodynamic and pharmacokinetic properties, and therapeutic potential in epilepsy. Drugs.1993, 45, (5), 760-787.
  2. W Gristwood; K Wilson. Kinetics of some benzothiazoles, benzoxazoles, and quinolines as substrates and inhibitors of rabbit liver aldehyde oxidase. Xenobiotica.1988, 18, (8), 949-954.