2-Cyclopenten-1-one, 98%
2-Cyclopenten-1-one, 98%
2-Cyclopenten-1-one, 98%
Thermo Scientific Chemicals

2-Cyclopenten-1-one, 98%

CAS: 930-30-3 | C5H6O | 82.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B23408.06
also known as B23408-06
Price (USD)
71.65
Online Exclusive
79.00
Save 7.35 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
71.65
Online Exclusive
79.00
Save 7.35 (9%)
Each
Chemical Identifiers
CAS930-30-3
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4795-1.4835 @ 20°C (non-U.S. specification)
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5% (non-U.S. specification)
View more
2-Cyclopenten-1-one is a versatile electrophile employed in various addition reactions including conjugate addition of organocopper nucleophiles, Michael reaction with silyl enol ethers and siloxanes. It is also used in Diels-Alder cycloadditions and phosphoniosilylations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyclopenten-1-one is a versatile electrophile employed in various addition reactions including conjugate addition of organocopper nucleophiles, Michael reaction with silyl enol ethers and siloxanes. It is also used in Diels-Alder cycloadditions and phosphoniosilylations.

Solubility
Miscible with alcohol and ether. Immiscible with water.

Notes
Hygroscopic. Store in cool place. Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
RUO – Research Use Only

General References:

  1. Zaras, A. M.; Thion, S.; Dagaut, P. Computational Kinetic Study for the Unimolecular Decomposition of Cyclopentanone. Int. J. Chem. Kinet. 2015, 47 (7), 439-446.
  2. Mante, O. D.; Rodriguez, J. A.; Senanayake, S. D.; Babu, S. P. Catalytic conversion of biomass pyrolysis vapors into hydrocarbon fuel precursors. Green Chem. 2015, 17 (4), 2362-2368.