4-Cumylphenol, 97%
4-Cumylphenol, 97%
4-Cumylphenol, 97%
Thermo Scientific Chemicals

4-Cumylphenol, 97%

CAS: 599-64-4 | C15H16O | 212.292 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1000 g
Catalog number B23410.30
also known as B23410-30
Price (USD)
72.65
Special offer
Online exclusive
Ends: 30-Jun-2026
85.10
Save 12.45 (15%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
72.65
Special offer
Online exclusive
Ends: 30-Jun-2026
85.10
Save 12.45 (15%)
Each
Specifications
Beilstein1870517
CAS599-64-4
ChEBICHEBI:35092
Chemical Name or Material4-Cumylphenol
Density1.115
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only