2-Amino-6-methylbenzothiazole, 99%
2-Amino-6-methylbenzothiazole, 99%
2-Amino-6-methylbenzothiazole, 99%
Thermo Scientific Chemicals

2-Amino-6-methylbenzothiazole, 99%

CAS: 2536-91-6 | C8H8N2S | 164.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B23422.22
also known as B23422-22
Price (USD)
107.65
Special offer
Online exclusive
Ends: 30-Jun-2026
127.00
Save 19.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
107.65
Special offer
Online exclusive
Ends: 30-Jun-2026
127.00
Save 19.35 (15%)
Each
Chemical Identifiers
CAS2536-91-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
Identification (FTIR)Conforms
Melting Point136.0-142.0?C
Assay (GC)≥98.5%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only