Aminomethanesulfonic acid, 97%
Aminomethanesulfonic acid, 97%
Aminomethanesulfonic acid, 97%
Thermo Scientific Chemicals

Aminomethanesulfonic acid, 97%

CAS: 13881-91-9 | CH5NO3S | 111.115 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B23441.22
also known as B23441-22
Price (USD)
85.65
Online Exclusive
94.50
Save 8.85 (9%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
85.65
Online Exclusive
94.50
Save 8.85 (9%)
Each
Chemical Identifiers
CAS13881-91-9
IUPAC Nameaminomethanesulfonic acid
Molecular FormulaCH5NO3S
InChI KeyOBESRABRARNZJB-UHFFFAOYSA-N
SMILESNCS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)>96.0%
Formpowder
Aminomethanesulfonic acid is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aminomethanesulfonic acid is used as a pharmaceutical intermediates.

Solubility
Soluble in water. (17 g/L) at 17°C

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents. Store at room temperature.
RUO – Research Use Only
John B. Lombardini. Opposite effects of 2-aminoethanesulfonic acid (taurine) and aminomethanesulfonic acid on calcium ion uptake in rat retinal preparations. European Journal of Pharmacology.1985, 110, 385-387.