4-Chloro-3-nitrobenzophenone, 99%
4-Chloro-3-nitrobenzophenone, 99%
4-Chloro-3-nitrobenzophenone, 99%
4-Chloro-3-nitrobenzophenone, 99%
Thermo Scientific Chemicals

4-Chloro-3-nitrobenzophenone, 99%

CAS: 56107-02-9 | C13H8ClNO3 | 261.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B23451.22
also known as B23451-22
Price (USD)
77.40
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
77.40
Each
Add to cart
Chemical Identifiers
CAS56107-02-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystals or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)103-108?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only