2-Methylbenzotrifluoride, 97%
2-Methylbenzotrifluoride, 97%
2-Methylbenzotrifluoride, 97%
Thermo Scientific Chemicals

2-Methylbenzotrifluoride, 97%

CAS: 13630-19-8 | C8H7F3 | 160.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B23453.06
also known as B23453-06
Price (USD)
168.65
Online Exclusive
187.00
Save 18.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
168.65
Online Exclusive
187.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS13630-19-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4320-1.4370 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only