2-Chloro-3',4'-dihydroxyacetophenone, 97%
2-Chloro-3',4'-dihydroxyacetophenone, 97%
2-Chloro-3',4'-dihydroxyacetophenone, 97%
Thermo Scientific Chemicals

2-Chloro-3',4'-dihydroxyacetophenone, 97%

CAS: 99-40-1 | C8H7ClO3 | 186.591 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B23496.22
also known as B23496-22
Price (USD)
143.65
Special offer
Online exclusive
Ends: 30-Jun-2026
169.00
Save 25.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
143.65
Special offer
Online exclusive
Ends: 30-Jun-2026
169.00
Save 25.35 (15%)
Each
Chemical Identifiers
CAS99-40-1
IUPAC Name2-chloro-1-(3,4-dihydroxyphenyl)ethan-1-one
Molecular FormulaC8H7ClO3
InChI KeyLWTJEJCZJFZKEL-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1O)C(=O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey or cream to brown or purple
Assay (HPLC)>96.0%
FormPowder
2-Chloro-3',4'-dihydroxyacetophenone is used as a styptic adrenobazone, quasi adrenaline drug gasp spirit of intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-3′,4′-dihydroxyacetophenone is used as a styptic adrenobazone, quasi adrenaline drug gasp spirit of intermediates.

Solubility
Soluble in DMSO, methanol.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only
Huidobro-Toro JP; Carpi A. Selective blocking properties of chloro acetyl catechol (CAC) on the alpha-adrenergic receptor. Europe PMC. 1976, 222 (2), 180-192.