3,5-Bis(trifluoromethyl)styrene, 96%, stab.
3,5-Bis(trifluoromethyl)styrene, 96%, stab.
3,5-Bis(trifluoromethyl)styrene, 96%, stab.
3,5-Bis(trifluoromethyl)styrene, 96%, stab.
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)styrene, 96%, stab.

CAS: 349-59-7 | C10H6F6 | 240.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B23499.06
also known as B23499-06
Price (USD)
337.65
Online Exclusive
375.00
Save 37.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
337.65
Online Exclusive
375.00
Save 37.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS349-59-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥92.0%
Refractive Index1.4235-1.4275 @ 20°C
Solution TestClear (10% in diethyl ether)
View more
3,5-Bis(trifluoromethyl)styrene is used to produce other chemicals for example 1-(3, 5-bis-Trifluoromethyl-phenyl)-ethane-1, 2-diol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Bis(trifluoromethyl)styrene is used to produce other chemicals for example 1-(3, 5-bis-Trifluoromethyl-phenyl)-ethane-1, 2-diol.

Solubility
Sparingly soluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only
Hongxiang Teng; Liping Lou; Kotaro Koike;Yasuhiro Koike; Yoshi Okamoto. Synthesis and characterization of trifluoromethyl substituted styrene polymers and copolymers with methacrylates: Effects of trifluoromethyl substituent on styrene. Polymer. 2011, 52, (4), 949-953.