3-Bromophenylhydrazine hydrochloride, 98%
3-Bromophenylhydrazine hydrochloride, 98%
3-Bromophenylhydrazine hydrochloride, 98%
Thermo Scientific Chemicals

3-Bromophenylhydrazine hydrochloride, 98%

CAS: 27246-81-7 | C6H8BrClN2 | 223.50 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B23510.14
also known as B23510-14
Price (USD)
300.65
Online Exclusive
334.00
Save 33.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
300.65
Online Exclusive
334.00
Save 33.35 (10%)
Each
Chemical Identifiers
CAS27246-81-7
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Appearance (Color)Pale cream to cream to brown or pale pink
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only