2-Amino-4-thiazoleacetic acid, 97%
2-Amino-4-thiazoleacetic acid, 97%
2-Amino-4-thiazoleacetic acid, 97%
Thermo Scientific Chemicals

2-Amino-4-thiazoleacetic acid, 97%

CAS: 29676-71-9 | C5H6N2O2S | 158.175 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B23534.14
also known as B23534-14
Price (USD)
49.65
Online Exclusive
55.30
Save 5.65 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
49.65
Online Exclusive
55.30
Save 5.65 (10%)
Each
Chemical Identifiers
CAS29676-71-9
IUPAC Name2-(2-amino-1,3-thiazol-4-yl)acetic acid
Molecular FormulaC5H6N2O2S
InChI KeyDYCLHZPOADTVKK-UHFFFAOYSA-N
SMILESNC1=NC(CC(O)=O)=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Appearance (Color)White to pale cream
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only