2,3,5-Collidine, 99%
2,3,5-Collidine, 99%
2,3,5-Collidine, 99%
Thermo Scientific Chemicals

2,3,5-Collidine, 99%

CAS: 695-98-7 | C8H11N | 121.183 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B23537.14
also known as B23537-14
Price (USD)
105.65
Online Exclusive
117.00
Save 11.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
105.65
Online Exclusive
117.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS695-98-7
IUPAC Name2,3,5-trimethylpyridine
Molecular FormulaC8H11N
InChI KeyGFYHSKONPJXCDE-UHFFFAOYSA-N
SMILESCC1=CN=C(C)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥98.5%
Refractive Index1.5065-1.5095 @ 20°C
FormLiquid
2,3,5-Collidine is omeprazole intermediate which is the drug involved in proton pump inhibitors to block the production of stomach acid. It is used in the manufacture vitamin D3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,5-Collidine is omeprazole intermediate which is the drug involved in proton pump inhibitors to block the production of stomach acid. It is used in the manufacture vitamin D3.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents. Store at room temperature.
RUO – Research Use Only
Mittelbach M.; Schmidt HW.; Uray G.; Junek H.; Lamm B.; Ankner K.; Brändström A.; Simonsson R. Synthesis of 4-methoxy-2,3,5-trimethylpyridine: a specific building block for compounds with gastric-acid inhibiting activity. Acta Chemica Scandinavica. Series B: Organic Chemistry and Biochemistry. 1988, 42, (8), 524-529.