3-Sulfobenzoic acid monosodium salt, 97%
3-Sulfobenzoic acid monosodium salt, 97%
3-Sulfobenzoic acid monosodium salt, 97%
Thermo Scientific Chemicals

3-Sulfobenzoic acid monosodium salt, 97%

CAS: 17625-03-5 | C7H5NaO5S | 224.162 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B23556.14
also known as B23556-14
Price (USD)
28.20
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
28.20
Each
Chemical Identifiers
CAS17625-03-5
IUPAC Namesodium 3-sulfobenzoate
Molecular FormulaC7H5NaO5S
InChI KeyKQHKITXZJDOIOD-UHFFFAOYSA-M
SMILES[Na+].OS(=O)(=O)C1=CC=CC(=C1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
FormPowder
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only