2-Chloro-4-fluorobenzyl bromide, 97%
2-Chloro-4-fluorobenzyl bromide, 97%
2-Chloro-4-fluorobenzyl bromide, 97%
Thermo Scientific Chemicals

2-Chloro-4-fluorobenzyl bromide, 97%

CAS: 45767-66-6 | C7H5BrClF | 223.469 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B23565.14
also known as B23565-14
Price (USD)
58.65
Online Exclusive
65.10
Save 6.45 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
58.65
Online Exclusive
65.10
Save 6.45 (10%)
Each
Chemical Identifiers
CAS45767-66-6
IUPAC Name1-(bromomethyl)-2-chloro-4-fluorobenzene
Molecular FormulaC7H5BrClF
InChI KeyGAUUDQVOPUKGJD-UHFFFAOYSA-N
SMILESFC1=CC=C(CBr)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormCrystals or fused solid
Melting Point (clear melt)33.0-42.0?C
Appearance (Color)Colorless or white
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only