8-Nitroquinoline, 98%
8-Nitroquinoline, 98%
8-Nitroquinoline, 98%
Thermo Scientific Chemicals

8-Nitroquinoline, 98%

CAS: 607-35-2 | C9H6N2O2 | 174.159 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B23574.14
also known as B23574-14
Price (USD)
127.65
Online exclusive
142.00
Save 14.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
127.65
Online exclusive
142.00
Save 14.35
Each
Specifications
Beilstein135178
CAS607-35-2
Chemical Name or Material8-Nitroquinoline
EINECS Number210-135-9
Formula Weight174.16
View more
8-Nitroquinoline is used in the preparation of bisubstituted phenoxy-6-methoxy-8-aminoquinoline. It is used to prepare furazano [3,4-h] quinoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Nitroquinoline is used in the preparation of bisubstituted phenoxy-6-methoxy-8-aminoquinoline. It is used to prepare furazano [3,4-h] quinoline.

Solubility
Soluble in ethanol, ethyl ether and benzene, slightly soluble in cold water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only

General References:

  1. M Hasegawa; T Takabatake; T Miyazawa. [Formations and reactions of aromatic furazan compounds]. YAKUGAKU ZASSHI . 2001, 121, (6), 379-393.
  2. C Chen; X Y Zheng; H Z Guo. [Studies on the tissue schizonticide of malaria parasite: synthesis of derivatives of 2-substituted phenoxyprimaquine, 4-methylprimaquine and quinoxaline]. Yao xue xue bao = Acta pharmaceutica Sinica. 1992, 27, (6), 418-422.