7-Fluorobenzofurazan-4-sulfonamide, 98%
7-Fluorobenzofurazan-4-sulfonamide, 98%
7-Fluorobenzofurazan-4-sulfonamide, 98%
Thermo Scientific Chemicals

7-Fluorobenzofurazan-4-sulfonamide, 98%

CAS: 91366-65-3 | C6H4FN3O3S | 217.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Catalog number B23586.MB
also known as B23586-MB
Price (USD)
391.65
Special offer
Online exclusive
Ends: 28-Dec-2026
460.00
Save 68.35
Each
Quantity:
25 mg
Request bulk or custom format
Price (USD)
391.65
Special offer
Online exclusive
Ends: 28-Dec-2026
460.00
Save 68.35
Each
Chemical Identifiers
CAS91366-65-3
IUPAC Name7-fluoro-2,1,3-benzoxadiazole-4-sulfonamide
Molecular FormulaC6H4FN3O3S
InChI KeyXROXHZMRDABMHS-UHFFFAOYSA-N
SMILESNS(=O)(=O)C1=CC=C(F)C2=NON=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
Melting Point (clear melt)142.0-149.0?C
FormPowder
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only