2-Chlorotoluene, 98%
2-Chlorotoluene, 98%
2-Chlorotoluene, 98%
Thermo Scientific Chemicals

2-Chlorotoluene, 98%

CAS: 95-49-8 | C7H7Cl | 126.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1000 g
Catalog number B23596.0B
also known as B23596-0B
Price (USD)
77.60
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
77.60
Each
Chemical Identifiers
CAS95-49-8
IUPAC Name1-chloro-2-methylbenzene
Molecular FormulaC7H7Cl
InChI KeyIBSQPLPBRSHTTG-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.5235-1.5265 @ 20?C
FormLiquid
Identification (FTIR)Conforms
2-Chlorotoluene is used for permanganate oxidation to give o-chlorobenzoic acid. It is used in the preparation of 3-chloro-4-methylcatechol and 4-chloro-3-methylcatechol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorotoluene is used for permanganate oxidation to give o-chlorobenzoic acid. It is used in the preparation of 3-chloro-4-methylcatechol and 4-chloro-3-methylcatechol.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only

General References:

  1. Daniel Dobslaw; Karl-Heinrich Engesser. Degradation of 2-chlorotoluene by Rhodococcus sp. OCT 10. Applied Microbiology and Biotechnology. 2012, 93, 2205-2214.
  2. Katrin Pollmann; Victor Wray and Dietmar H. Pieper. Chloromethylmuconolactones as Critical Metabolites in the Degradation of Chloromethylcatechols: Recalcitrance of 2-Chlorotoluene. Journal of Bacteriology. 2005, 187, (7), 2332-2340.
  3. For permanganate oxidation to give o-chlorobenzoic acid, see: Org. Synth. Coll., 2, 135 (1943).