4-Amino-2,5-difluorobenzonitrile, 96%
4-Amino-2,5-difluorobenzonitrile, 96%
4-Amino-2,5-difluorobenzonitrile, 96%
Thermo Scientific Chemicals

4-Amino-2,5-difluorobenzonitrile, 96%

CAS: 112279-61-5 | C7H4F2N2 | 154.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B23611.03
also known as B23611-03
Price (USD)
33.50
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
33.50
Each
Specifications
CAS112279-61-5
Chemical Name or Material4-Amino-2,5-difluorobenzonitrile
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Formula Weight154.12
Health Hazard 1H301-H312+H332-H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only