Copper(II) acetate, anhydrous, 98%
Copper(II) acetate, anhydrous, 98%
Copper(II) acetate, anhydrous, 98%
Thermo Scientific Chemicals

Copper(II) acetate, anhydrous, 98%

CAS: 142-71-2 | C4H6CuO4 | 181.634 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B23615.36
also known as B23615-36
Price (USD)
597.65
Online Exclusive
664.00
Save 66.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
597.65
Online Exclusive
664.00
Save 66.35 (10%)
Each
Chemical Identifiers
CAS142-71-2
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>97.5%
pH5.0-5.3
Formpowder
Elemental AnalysisPb:<20ppm
Appearance (Color)Blue-green
View more
Copper(II) acetate is used in biochemical applications such as DNA extraction. It is used as a source of copper in inorganic synthesis, an oxidizing agent and as a catalyst in organic synthesis. It is also used to couple two terminal alkynes to make a 1,3-diyne. It is widely used in pigments, manufacture of paris green, in textile dyeing, skin conditioner in cosmetics, and as antioxidant and stabilizer in food grade polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) acetate is used in biochemical applications such as DNA extraction. It is used as a source of copper in inorganic synthesis, an oxidizing agent and as a catalyst in organic synthesis. It is also used to couple two terminal alkynes to make a 1,3-diyne. It is widely used in pigments, manufacture of paris green, in textile dyeing, skin conditioner in cosmetics, and as antioxidant and stabilizer in food grade polymers.

Solubility
Slightly soluble in water. Soluble in alcohol and ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Mansour, A. M. What happens when (1H-benzimidazol-2-ylmethyl)-N-phenyl amine is added to copper(II) acetate? Spectroscopic, magnetic, and DFT studies. Inorg. Chim. Acta. 2013, 408, 186-192.
  2. Abdalla, K.; Rahmat, A.; Azizan, A. Effect of copper (II) acetate pretreatment on zinc phosphate coating morphology and corrosion resistance. J. Coat. Technol. Res. 2013, 10 (1), 133-139.