Thiophene-3-boronic acid, 98%
Thiophene-3-boronic acid, 98%
Thiophene-3-boronic acid, 98%
Thiophene-3-boronic acid, 98%
Thermo Scientific Chemicals

Thiophene-3-boronic acid, 98%

CAS: 6165-69-1 | C4H5BO2S | 127.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B23637.14
also known as B23637-14
Price (USD)
339.65
Online Exclusive
377.00
Save 37.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
339.65
Online Exclusive
377.00
Save 37.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS6165-69-1
IUPAC Name(thiophen-3-yl)boronic acid
Molecular FormulaC4H5BO2S
InChI KeyQNMBSXGYAQZCTN-UHFFFAOYSA-N
SMILESOB(O)C1=CSC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥97.5%
Proton NMRConforms to structure
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only