1-Iodo-2,4-dinitrobenzene, 98%
1-Iodo-2,4-dinitrobenzene, 98%
1-Iodo-2,4-dinitrobenzene, 98%
Thermo Scientific Chemicals

1-Iodo-2,4-dinitrobenzene, 98%

CAS: 709-49-9 | C6H3IN2O4 | 294.004 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B23659.14
also known as B23659-14
Price (USD)
272.65
Special offer
Online exclusive
Ends: 30-Jun-2026
321.00
Save 48.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
272.65
Special offer
Online exclusive
Ends: 30-Jun-2026
321.00
Save 48.35 (15%)
Each
Chemical Identifiers
CAS709-49-9
IUPAC Name1-iodo-2,4-dinitrobenzene
Molecular FormulaC6H3IN2O4
InChI KeyFXMKXMJLXRTQSW-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(I)C(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)85.0-91.0?C
Appearance (Color)Yellow to orange to brown
FormPowder
Assay (GC)≥97.5%
It is employed in the cloning and characterization of a biotic-stress-inducible glutathione transferase from Phaseolus vulgaris.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the cloning and characterization of a biotic-stress-inducible glutathione transferase from Phaseolus vulgaris.

Solubility
Insoluble in water.

Notes
Light sensitive. Store away from oxidizing agents, bases and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under dark.
RUO – Research Use Only

General References:

  1. J. Burnett, et al. Notes - Improved Preparation of 1-Iodo-2,4-dinitrobenzene.J. Org. Chem.,1958,23(2), 305-306.
  2. Christiane Schrenk, et. al. GlutathioneS-Transferase Activity in Aquatic Macrophytes with Emphasis on Habitat Dependence.Ecotoxicology and Environmental Safety.,1998,40(3), 226-233.