2-Phenylindole, 95%
2-Phenylindole, 95%
2-Phenylindole, 95%
Thermo Scientific Chemicals

2-Phenylindole, 95%

CAS: 948-65-2 | C14H11N | 193.249 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number B23674.18
also known as B23674-18
Price (USD)
27.65
Special offer
Online exclusive
Ends: 28-Dec-2026
32.70
Save 5.05
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
27.65
Special offer
Online exclusive
Ends: 28-Dec-2026
32.70
Save 5.05
Each
Chemical Identifiers
CAS948-65-2
IUPAC Name2-phenyl-1H-indole
Molecular FormulaC14H11N
InChI KeyKLLLJCACIRKBDT-UHFFFAOYSA-N
SMILESN1C2=CC=CC=C2C=C1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥94.0%
Melting Point (clear melt)186.5-195.5?C
Appearance (Color)White to cream to green to pale brown
CommentMay contain a small quantity of fine particulate matter from the manufacturing process.
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only