Biphenyl-4-boronic acid, 97+%
Biphenyl-4-boronic acid, 97+%
Biphenyl-4-boronic acid, 97+%
Thermo Scientific Chemicals

Biphenyl-4-boronic acid, 97+%

CAS: 5122-94-1 | C12H11BO2 | 198.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B23703.03
also known as B23703-03
Price (USD)
33.65
Online exclusive
37.20
Save 3.55
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
33.65
Online exclusive
37.20
Save 3.55
Each
Chemical Identifiers
CAS5122-94-1
IUPAC Name{[1,1'-biphenyl]-4-yl}boronic acid
Molecular FormulaC12H11BO2
InChI KeyXPEIJWZLPWNNOK-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMRConforms to structure
Assay (HPLC)≥98.0%
Appearance (Color)White to cream
FormPowder
Assay (Aqueous acid-base Titration)≥98.0%
Used in Suzuki reactions and as intermediates of liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in Suzuki reactions and as intermediates of liquid crystals.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Michael S. Malamas, et al. Novel Benzofuran and Benzothiophene Biphenyls as Inhibitors of Protein Tyrosine Phosphatase 1B with Antihyperglycemic Properties.J. Med. Chem.,2000,43(7), 1293-1310.
  2. Soon Ok Jeon, et al. Spiro[fluorene-7,9'-benzofluorene] host and dopant materials for blue light-emitting electroluminescence device.Synth. Met.,2009,159(12), 1147-1152.
  3. The absolute configuration of 1-arylethane-1,2-diols has been determined by analysis of the CD spectra of the 4-biphenylboronates: Org. Lett., 1, 2093 (1999).