2-Methyl-4-(trifluoromethoxy)aniline, 97+%
2-Methyl-4-(trifluoromethoxy)aniline, 97+%
2-Methyl-4-(trifluoromethoxy)aniline, 97+%
Thermo Scientific Chemicals

2-Methyl-4-(trifluoromethoxy)aniline, 97+%

CAS: 86256-59-9 | C8H8F3NO | 191.153 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B23706.06
also known as B23706-06
Price (USD)
99.65
Online Exclusive
110.00
Save 10.35 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
99.65
Online Exclusive
110.00
Save 10.35 (9%)
Each
Chemical Identifiers
CAS86256-59-9
IUPAC Name2-methyl-4-(trifluoromethoxy)aniline
Molecular FormulaC8H8F3NO
InChI KeyIIDBMILLZRYZCH-UHFFFAOYSA-N
SMILESCC1=CC(OC(F)(F)F)=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4660-1.4710 @ 20?C
Assay (GC)≥97.0%
Appearance (Color)Clear pale yellow to pale orange
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only