Manganese(II) acetate, anhydrous, 98+%
Manganese(II) acetate, anhydrous, 98+%
Manganese(II) acetate, anhydrous, 98+%
Thermo Scientific Chemicals

Manganese(II) acetate, anhydrous, 98+%

CAS: 638-38-0 | C4H6MnO4 | 173.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number B23708.30
also known as B23708-30
Price (USD)
205.00
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
205.00
Each
Chemical Identifiers
CAS638-38-0
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Complexometric Titration≥98.0 to ≤102.0%
Appearance (Color)White to pink or grey to pale brown
Manganese(II) acetate is used as a dye mordant and varnish drier. It is also used in fertilizers, food packaging and feed additives. Further, it serves as a desiccant, catalyst, intermediate and ion exchange agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(II) acetate is used as a dye mordant and varnish drier. It is also used in fertilizers, food packaging and feed additives. Further, it serves as a desiccant, catalyst, intermediate and ion exchange agent.

Solubility
Soluble in water, methanol and acetic acid.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong bases and strong acids.
RUO – Research Use Only

General References:

  1. Naeem, R.; Yahya, R.; Pandikumar, A.; Huang, N. M.; Misran, M.; Arifin, Z.; Mazhar, M. Photoelectrochemical properties of morphology controlled manganese, iron, nickel and copper oxides nanoball thin films deposited by electric field directed aerosol assisted chemical vapour deposition. Mater. Today Commun. 2015, 4, 141-148.
  2. Arun, N.; Aravindan, V.; Ling, W. C.; Madhavi, S. Importance of nanostructure for reversible Li-insertion into octahedral sites of LiNi0.5Mn1.5O4 and its application towards aqueous Li-ion chemistry. J. Power Sources 2015, 280, 240-245.