Ethanesulfonyl chloride, 98+%
Ethanesulfonyl chloride, 98+%
Ethanesulfonyl chloride, 98+%
Ethanesulfonyl chloride, 98+%
Thermo Scientific Chemicals

Ethanesulfonyl chloride, 98+%

CAS: 594-44-5 | C2H5ClO2S | 128.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B23723.36
also known as B23723-36
Price (USD)
214.65
Online Exclusive
238.00
Save 23.35 (10%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
214.65
Online Exclusive
238.00
Save 23.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS594-44-5
IUPAC Nameethanesulfonyl chloride
Molecular FormulaC2H5ClO2S
InChI KeyFRYHCSODNHYDPU-UHFFFAOYSA-N
SMILESCCS(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4510-1.4550 @ 20?C
Appearance (Color)Clear colorless to pale yellow or purple
FormLiquid
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only