(3-Mercaptopropyl)trimethoxysilane, 95%
(3-Mercaptopropyl)trimethoxysilane, 95%
(3-Mercaptopropyl)trimethoxysilane, 95%
Thermo Scientific Chemicals

(3-Mercaptopropyl)trimethoxysilane, 95%

CAS: 4420-74-0 | C6H16O3SSi | 196.336 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B23726.36
also known as B23726-36
Price (USD)
259.65
Online Exclusive
288.00
Save 28.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
259.65
Online Exclusive
288.00
Save 28.35 (10%)
Each
Chemical Identifiers
CAS4420-74-0
IUPAC Name3-(trimethoxysilyl)propane-1-thiol
Molecular FormulaC6H16O3SSi
InChI KeyUUEWCQRISZBELL-UHFFFAOYSA-N
SMILESCO[Si](CCCS)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥94.0%
Refractive Index1.4405-1.4465 @ 20?C (UK Sourced Material)
CommentMaterial sourced in UK and US
View more
(3-Mercaptopropyl)trimethoxysilane is used as a capping molecule for cadmium sulphide nanocrystals. It is used in the preparation of multi-walled carbon nanotubes and bilayer modified gold electrode. It acts as a linker molecule used in the preparation of gold multilayers and as a molecular adhesive in the fabrication of vapor-deposited gold electrodes on glass substrates. It is also used as a precursor for silica.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Mercaptopropyl)trimethoxysilane is used as a capping molecule for cadmium sulphide nanocrystals. It is used in the preparation of multi-walled carbon nanotubes and bilayer modified gold electrode. It acts as a linker molecule used in the preparation of gold multilayers and as a molecular adhesive in the fabrication of vapor-deposited gold electrodes on glass substrates. It is also used as a precursor for silica.

Solubility
Miscible with methanol, ethanol, isopropanol, acetone, benzene, toluene and xylene.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, strong acids, alcohols and water. Decomposes in water.
RUO – Research Use Only

General References:

  1. Hau, N. Y.; Chang, Y. H.; Feng, S. P. Kinetics Study of Silver Electrocrystallization on (3-mercaptopropyl)trimethoxysilane-grafted Indium Tin Oxide Plastic Substrate. Electrochim. Acta 2015, 158, 121-128.
  2. Mosa, J.; Durán, A.; Aparicio, M. Sulfonic acid-functionalized hybrid organic-inorganic proton exchange membranes synthesized by sol-gel using 3-mercaptopropyl trimethoxysilane (MPTMS). J. Power Sources 2015, 297, 208-216.