n-Octyltrichlorosilane, 97%
n-Octyltrichlorosilane, 97%
n-Octyltrichlorosilane, 97%
Thermo Scientific Chemicals

n-Octyltrichlorosilane, 97%

CAS: 5283-66-9 | C8H17Cl3Si | 247.659 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B23753.14
also known as B23753-14
Price (USD)
29.65
Special offer
Online exclusive
Ends: 30-Jun-2026
35.20
Save 5.55 (16%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
29.65
Special offer
Online exclusive
Ends: 30-Jun-2026
35.20
Save 5.55 (16%)
Each
Chemical Identifiers
CAS5283-66-9
IUPAC Nametrichloro(octyl)silane
Molecular FormulaC8H17Cl3Si
InChI KeyRCHUVCPBWWSUMC-UHFFFAOYSA-N
SMILESCCCCCCCC[Si](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow or pale brown
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.4460-1.4510 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only