Diethyl diethylmalonate, 98%
Diethyl diethylmalonate, 98%
Diethyl diethylmalonate, 98%
Thermo Scientific Chemicals

Diethyl diethylmalonate, 98%

CAS: 77-25-8 | C11H20O4 | 216.277 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B23810.14
also known as B23810-14
Price (USD)
43.65
Online Exclusive
48.60
Save 4.95 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
43.65
Online Exclusive
48.60
Save 4.95 (10%)
Each
Chemical Identifiers
CAS77-25-8
IUPAC Name1,3-diethyl 2,2-diethylpropanedioate
Molecular FormulaC11H20O4
InChI KeyZKBBUZRGPULIRN-UHFFFAOYSA-N
SMILESCCOC(=O)C(CC)(CC)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Clear colorless
Refractive Index1.4220-1.4260 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only